Safety wash bottles, wide neck, multilingual, Azlon®
LDPE, translucent, with PP spray cap.
- Indelibly printed in four languages (English, French, German and Spanish)
- Choose from three water or five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
- Closure feature precision jet tip aperture for fine control
Wide mouth for easy, safe filling. Fitted with colour coded one-piece PP closure that maintain their shape in use.
: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.