Perchloric acid ≥70% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Merck Index:14,7153
Boiling Pt:203 °C
Melting Pt:–18 °C
Density:1.670
Flash Pt:113 °C (235 °F)
Storage Temperature:Ambient
EINECS:231-512-4
UN:1873
ADR:5.1,I
:GHS03
:GHS05