Order Entry
Finland
ContactUsLinkComponent
Hydrochloric acid 36.5 - 38.0%, GenAR® NF, Macron Fine Chemicals™
Corrosive
Irritant
Hydrochloric acid 36.5 - 38.0%, GenAR® NF, Macron Fine Chemicals™
Catalog # C2626-46
CAS Number:  
undefined
Corrosive
Irritant
Hydrochloric acid 36.5 - 38.0%, GenAR® NF, Macron Fine Chemicals™
Catalog # C2626-46
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, clear
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • GMP Manufactured Product:
  • Meets E.P. Chemical Specifications:
  • Meets B.P. Chemical Specifications:
  • Meets J.P. Chemical Specifications:
  • Meets N.F. Requirements:
  • Store below 25°C:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Residue on Ignition:
    ≤ 0.008 %
  • NF - Bromide or Iodide:
    Passes Test
  • NF - Free Bromine or Chlorine:
    Passes Test
  • NF - Sulfate (SO₄):
    Passes Test
  • NF - Sulfite (SO₃):
    Passes Test
  • NF - Assay (HCl)(NF):
    36.5 - 38.0 %
  • Identification A (EP/BP):
    Passes Test
  • Identification B (EP/BP):
    Passes Test
  • Identification C (EP/BP):
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Free Chlorine (as Cl):
    ≤ 4 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 20 ppm
  • EP/BP - Residue after Evaporation:
    ≤ 0.01 %
  • Assay (HCl)(EP/BP):
    35.0 - 39.0 %
  • JP - Assay (HCl)(JP):
    35.0 - 38.0 %
  • JP - Identification:
    Passes Test
  • JP - Residue on ignition, mg/10mL:
    ≤ 1.0
  • JP - Bromide or Iodide:
    Passes Test
  • JP - Free Bromine or Chlorine:
    Passes Test
  • JP - Sulfate (SO₄):
    Passes Test
  • JP - Sulfite (SO₃):
    Passes Test
  • JP - Arsenic (As):
    ≤ 1 ppm
  • JP - Heavy Metals (as Pb) (JP):
    ≤ 5 ppm
  • JP - Mercury (Hg):
    ≤ 0.04 ppm

Specification Test Results

About this item

 :
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–35 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
 :Management of Change (MOC) category = N
 :36.46 g/mol
EINECS:231-595-7
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324