

1,2-Dichlorobenzene-[D₄] (98% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2199-69-1
:
Formula:C₆D₄Cl₂
MDL Number:MFCD00037106
Boiling Pt:178 °C (1013 hPa)
Melting Pt:-17 °C
Density:1.34 g/cm³
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:218-606-0
ADR:6.1,III
:150.97 g/mol
UN:1591
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.2 % (Coulometric)
- D-Enrichment:≥97.5 %