Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Rekisteröidy
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dimethoxytetrahydrofuran
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture ≥98%
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture ≥98%
:
Thermo Fisher Scientific
2,5-Dimethoxytetrahydrofuran cis- and trans-mixture ≥98%
:
Thermo Fisher Scientific
:
696-59-3
:
:
C₆H₁₂O₃
:
MFCD00005359
:
132.16 g/mol
:
145…147 °C
:
–45 °C
:
1.020
:
35 °C (95 °F)
:
Ambient
:
211-797-1
:
3271
:
3,III
:
GHS02
:
GHS07
msds_ALFAA11687
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years