Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Barium
Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)
Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)
:
Thermo Fisher Scientific
Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)
:
Thermo Fisher Scientific
:
7440-39-3
:
:
Ba
:
MFCD00134031
:
137.33 g/mol
:
12,00991
:
1640 °C (1013 hPa)
:
725 °C
:
3.51 g/cm³ (20 °C)
:
Ambient
:
231-149-1
:
1400
:
4.3,II
:
GHS02
msds_ALFA000653
Thermo Scientific Acroseal 25 Years