Powder funnels with short, wide stem
Supplier: Brand
Certificates
About this item
Borosilicate glass 3,3.
- Chemically resistant glass
- Minimal thermal expansion, giving relatively high resistance to temperature changes
- Quick and easy to clean
Ideal for transferring powders, can also be used for liquids.