Order Entry
Finland
Orders LinkContactUsLinkComponent
2-Propanol

2-Propanol

Glass bottle, 2,5 L
Catalog # REGERH1018
Flammable
Irritant
Supplier:   REAGECON
2-Propanol
Catalog # REGERH1018
Supplier:  REAGECON
CAS Number:  67-63-0
2-Propanol
2-Propanol

Glass bottle, 2,5 L

CAS Number:  67-63-0
Price
156,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.