Xylene cyanole FF, Ultrapure
Catalog # 0819-20G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:20 g
- Seq:3290
Specifications
Specification Test Results
- DNase:none detected
- Em (614 nm, Water):≥30000
- Lambda Max:612 – 616 nm
- Loss on Drying:≤10%
- RNase:none
Specification Test Results
About this item
:
:538.62 g/mol
Melting Pt:295 °C
EINECS:220-167-5
:GHS07
Storage Temperature:Ambient
CAS Number:2650-17-1
Formula:C₂₅H₂₇N₂NaO₆S₂
MDL Number:MFCD00040657