CAPS (3-(cyclohexylamino)propanesulphonic acid), Millipore®
Catalog # 239782-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Merck Index:13,01773
Melting Pt:324 °C
:221.32 g/mol
Storage Temperature:Ambient
EINECS:214-492-1
CAS Number:1135-40-6
Formula:C₆H₁₁NH(CH₂)₃SO₃H
MDL Number:MFCD00003837