Order Entry
Finland
ContactUsLinkComponent
(1S)-(-)-β-Pinene, Supelco®
(1S)-(-)-β-Pinene, Supelco®
Catalog # 80609-5ML
Supplier:  Merck
CAS Number:  
undefined
(1S)-(-)-β-Pinene, Supelco®
Catalog # 80609-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :136.24 g/mol
Merck Index:12,07600
Boiling Pt:165 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.9034 g/cm³ (20 °C)
Flash Pt:36 °C
EINECS:242-060-2
UN:2319
ADR:3,III
 :M-Clarity™ quality level = MQ100
CAS Number:18172-67-3
Formula:C₁₀H₁₆
MDL Number:MFCD00001345