Order Entry
Finland
ContactUsLinkComponent
Gelatin-Sepharose® 4B
Gelatin-Sepharose® 4B
Catalog # 17-0956-01
Supplier:  Cytiva
CAS Number:  
undefined
Gelatin-Sepharose® 4B
Catalog # 17-0956-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    45 - 165 µm
  • Pk:
    25 ml
  • Description:
    Gelatin-Sepharose® 4B
  • Matrix:
    4% agarose

Specifications

About this item

Gelatin Sepharose® 4B affinity media with immobilised gelatin is designed for the purification or depletion of fibronectin as gelatin binds to fibronectin with high specificity. Gelatin Sepharose® 4B is bovine gelatin coupled to Sepharose® 4B by the cyanogen bromide method.

Gelatin binds specifically to fibronectin, which is a high molecular weight glycoprotein found on the surfaces of many cell types and present in many extracellular fluids including plasma. Gelatin Sepharose 4B has been designed for the purification or removal of fibronectin.