Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Litmus
Litmus
Litmus
Litmus
Supplier:
CHROMA
Certificates
Expand image
Litmus
Supplier:
CHROMA
About this item
CAS Number:
1393-92-6
:
MDL Number:
MFCD00131528
EINECS:
215-739-6