Giemsa's stain, Technical Grade used as a blood stain, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:51811-82-6
:
MDL Number:MFCD00081642
Melting Pt:300 °C (lit.)
:M-Clarity™ quality level = MQ100
EINECS:257-438-2
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Color):Dark to Very Dark Green and Black
- Appearance (Form):Powder
- Solubility (Color):4 mg plus 4 ml of Ethanol
- Wavelength (1) (UV):645 - 655 nm
- Solvent (UV):Methanol