Bismarck brown Y, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:10114-58-6
:
Formula:C₁₈H₁₈N8·2HCl
MDL Number:MFCD00012977
:419.32 g/mol
Merck Index:12,01294
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:233-314-3