Order Entry
Finland
ContactUsLinkComponent
2,6-Dichlorophenolindophenol sodium salt hydrate 98+%
2,6-Dichlorophenolindophenol sodium salt hydrate 98+%
Certificates
undefined
2,6-Dichlorophenolindophenol sodium salt hydrate 98+%
Certificates
Specification Test Results
  • Appearance:
    Dark green to dark blue-purple Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤14 % (1 g, 120°C)
  • Titration with HClO4:
    ≥98.0 % (On dry substance)
  • Suitability:
    (for Ascorbic acid det.) Passes test