Order Entry
Finland
ContactUsLinkComponent
Carbol fuchsin, Sigma-Aldrich®
Carbol fuchsin, Sigma-Aldrich®
Certificates
undefined
Carbol fuchsin, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Green to Brown
    Conforms to Requirements
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Solubility (Colour)
    Dark Pink to Red
    c = 1 mg/ml; Ethanol:
    Conforms
  • Recommended Retest Period:
    4 Years

Specification Test Results