VWR®, Stirring shafts
Supplier: VWR Collection
Certificates
About this item
PTFE. For use with stirring blades. Chemically inert. The smooth surface reduces contamination and makes cleaning easier. It also prevents the agitation vessels from being scratched.
- Excellent chemical resistance
- Temperature resistant from −200 to 280 °C
Ordering information: Stirring blades must be ordered separately.