Order Entry
Finland
ContactUsLinkComponent
Orbital shakers, Vibramax
Orbital shakers, Vibramax
Certificates
undefined
Orbital shakers, Vibramax
Specifications
  • Catalog No:
  • 444-1220
  • Model:
  • Vibramax 100
  • Speed:
  • 150 - 2500 min⁻¹
  • Shaking motion:
  • Vibrating orbital
  • Max. load:
  • 2 kg
  • Power input/output:
  • 31/7 W
  • Orbit:
  • 3 mm
  • Weight:
  • 5,5 kg
  • W×D×H:
  • 245×310×125 mm
Specifications