Order Entry
Finland
ContactUsLinkComponent
Colour hygiene test, HY-RiSE®
Colour hygiene test, HY-RiSE®
Catalog # 1.31200.0001
Supplier:  Merck
undefined
Colour hygiene test, HY-RiSE®
Catalog # 1.31200.0001
Supplier:  Merck
Specifications
  • Description:
    HY-RiSE® (50 tests)
  • Pk:
    50
Specifications
About this item
The HY-RiSE® kit contains a colour hygiene test strip for manual assessment of the general cleanliness of surfaces, especially in the food and beverage industry. Detects food residues and microorganisms, so-called invisible dirt, which can be hazardous to health. The principle for testing is based on detection of NAD/NADH.
  • Much more sensitive than simple visual inspection
  • Easy to perform and read
  • Clear result within five minutes
  • No instrument required, all you need in a pocket format