Lift pumps for solvents
Supplier: NRG DEVELOPMENTS
Certificates
About this item
These lift pumps feature plastic sleeved steel piston rods so that only compatible materials comes in to contact with the fluid being handled. The standard adapter will fit for 2" BSB or 2" NPT threads. Nylon, with PTFE seals.
- Fitted with PTFE seals to offer maximum chemical resistance
- There are models to fit from 20 to 210 litre containers