Cleaning and disinfection machines, G 7883
Supplier: MIELE
Certificates
About this item
The G 7883 cleaner/disinfector is suitable for reprocessing laboratory glassware for analytical experiments in eg. industry, environmental and research laboratories. This appliance has a large two-level wash cabinet and can be integrated into a continuous worktop as a built-under unit or installed as a freestanding unit. After an intensive main cleaning phase, the disinfection takes place at >90 °C and a holding time of 5 minutes. The final rinsing is carried out with deionised water without rinsing agents.
- 10 standard cleaning programmes
- Main wash/rinse cycle optionally with fully demineralised water
- Thermal disinfection
- Electrical door lock
- With water softener inside and steam condenser
- Programme safety cut-out
- Optical and acoustic signal at end of programme
- Hygienic freshwater system with fresh water intake for each programme stage
- 3 spray arms for thorough cleaning of laboratory glassware
- Thorough cleaning of lumens with injector system
- Flowmeter to monitor water intake quantities
Greater capacity - Increased purity - More flexibility.
Certifications: In accordance with VDE, EMC/radio interference protection, DVGW, IP X1.
Ordering information: Baskets must be ordered separately. Other models, e.g. with cladding in white or with decorative border, are available on request. For usage as a freestanding unit, the cover must be ordered separately, please contact Avantor for details.