Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for proLab meters
Accessories for proLab meters
Accessories for proLab meters
Supplier:
SI Analytics
Certificates
Expand image
Accessories for proLab meters
Supplier:
SI Analytics
About this item