Weighing papers, grade 2122, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
These Grade 2122 weigh paper sheets reliably allow samples to be transferred to scales without adding unwanted substances that could impact analytical results.
- Smooth surface allows samples to slide off with ease and not be retained on paper
- Shiny pergamyne paper allows visual determination that all sample has been removed
- Extremely thin (0,03 mm) and lightweight (40 g/m²) for safe and secure sample transport
- No glue or additives (residue-free)
This paper is not low in nitrogen. Therefore, it should not be used for Kjeldahl analysis.