Chromatography tubing
Supplier: VICI JOUR RESEARCH
Certificates
About this item
Tefzel.
Tefzel tubing has a higher burst pressure than PTFE.
* Recommended max. permanent working pressure
Tefzel.
Tefzel tubing has a higher burst pressure than PTFE.
* Recommended max. permanent working pressure