Test tubes with SVL® screwcaps
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass, phenolic screwcaps.
- Excellent chemical and thermal resistance
- Screw caps with inert PTFE lined rubber seal
- Cap can withstand repeated dry heat and steam sterilisation cycles (121 °C for 2 hours)
Culture tubes with neck and SVL® standard screw neck.