Order Entry
Finland
ContactUsLinkComponent
VWR®, Culture and Test Tubes, Disposable
VWR®, Culture and Test Tubes, Disposable
Catalog # 211-0046
Supplier:  VWR Collection
   Low Manufacturing Impact
   Sustainable Packaging
undefined
VWR®, Culture and Test Tubes, Disposable
Catalog # 211-0046
Supplier:  VWR Collection
   Low Manufacturing Impact
   Sustainable Packaging

Specifications

  • Ext.Ø:
    12 mm
  • Packed:
    3000/Bag
  • Capacity:
    5 ml
  • Colour:
    Clear
  • Length:
    75 mm
  • Material:
    PS
  • Seq:
    11
  • Environmentally Preferable:
    Low Manufacturing Impact,
    Sustainable Packaging
  • Pk:
    3.000

Specifications

About this item

Transparent PS or translucent PP.

  • PS tubes: Not autoclavable, temperature resistant from –40 to +90 °C, can be centrifuged up to 1500×g
  • PP tubes: Autoclavable up to 121 °C, temperature resistant from –90 to +135 °C, can be centrifuged up to 3000×g
  • Press down stoppers or dual position closures available for secure sealing
  • Sustainable design based on virgin resins

Precision moulded reagent and sample tubes for a wide variety of applications, with conical or round bottom, sterile or non sterile.

Caps for culture and test tubes

Product Information