Tubes, rack packed, non-sterile
Supplier: Brand
Certificates
About this item
PP tubes and PE caps, non-sterile.
- Tubes and racks are autoclavable at 121 °C (2 bar) according to DIN EN 285 (not caps)
- Designed for storing and freezing down to −80 °C and for transporting reagents and samples
- Compatible with standard microplates
- Suitable for working with multi-channel pipettors
Suitable for biological tests, such as PCR, cell uptake studies, RIA and EIA.
Tube o. Ø×h: 8,8×45 mm