Order Entry
Finland
ContactUsLinkComponent
Microcentrifuge tubes, 1,5 ml
Microcentrifuge tubes, 1,5 ml
Catalog # 211-2610
Supplier:  Brand
undefined
Microcentrifuge tubes, 1,5 ml
Catalog # 211-2610
Supplier:  Brand
Specifications
  • Type:
    With attached lid
  • Version:
    Graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Colour:
    Colourless
  • Pk:
    500
Specifications
About this item
PP, graduated (only 211-2610 and 212-0198), autoclavable with attached cap (except 211-2611).
  • Can withstand centrifugation up to 20000×g (except 211-2611: Up to 6000×g)
  • Flat cap can be easily pierced
  • Frosted marking area (only 211-2610 and 212-0198)
  • Amber coloured microcentrifuge tubes are ideal for light-sensitive reagents
IVD (except 211-2611): CE-marked according to IVD-Directive 98/79 EC
Certifications: CE marked (only 211-2610 and 212-0198) according to IVD Directive 98/79 EC.