Immunology tubes
PS, clear. Star bottomed tubes have an increased surface area and thus make it possible to bind larger amounts of antigens or antibodies.
Immuno tubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone).
Available with either MICROLON® 200 (medium binding) and MICROLON® 600 (high binding) surfaces.
Ø×L: 12×75 mm
- Catalog No:
- 738-0002
- 738-0003
- 738-0000
- 738-0001
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)