Sealing cap assemblies, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Tefzel™ FEP plug, PP screw closure, Viton™ fluoroelastomer O-ring.
- PP screw closure, ETFE plug, Viton™ fluoroelastomer O-ring
- Top fits securely for use at maximum speeds without leakage
- Autoclavable/biohazard/leakproof
Process tubes at their highest rated speed level (10000×g or greater) without leakage, even when full. Sealing cap assemblies include a Viton™ fluoroelastomer O-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.
Specifications
- Catalog No:
- 525-2915
- 525-2914
- 525-2913
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement