Transport trolleys
Chrome plated steel frame with HDPE trough shaped trays.
- Stable, tough and versatile, easy to clean
- Load capacity: Max. 65 kg for each tray; combined 180 kg
- Rubber wheels to protect floors
The troughs are 70 mm deep, enabling liquids to be transported safely retaining any spillage. The height of the middle tray can be adjusted. Available in two sizes, with two or three trays.