Order Entry
Finland
ContactUsLinkComponent
Beaker tongs, stainless steel
Beaker tongs, stainless steel
  231-1009
 :  BOCHEM
undefined
Beaker tongs, stainless steel
  231-1009
 :  BOCHEM

 

  • Length:
    300 mm
  • Pk:
    1
  • Spanning area:
    60 - 100 mm

 

 

Stainless steel ¹⁸/₁₀, antimagnetic and electrolytical polished.

  • Jaws are insulated with ceramic
  • For beakers 100 to 1500 ml