Bench mats for cleanrooms
: DYCEM
This flexible bench mat can be placed on all kind of surfaces. The closed cell structure prevents the movement and loss of small components used in laboratories to stabilise fragile equipment. Approved for use in all classes of cleanrooms, including microelectronics, micro mechanics, semi-conductor, pharmaceutical, biotech and medical devices.
- Non-slip and anti-static properties
- Good shock absorption and stabilisation of sensitive equipment
- Improves accuracy, precision and assembly time
- Non-particulating: Helmke drum test
- Durable and easy to clean
- Portable, easy to transport
Non toxic material, heat resistant; resistant to most chemicals
Co-efficient of friction (µ): 3,5 (using Tortus floor friction tester)
Foot Traffic Only, expected life: 3 to 4 Years, warranty for 2 years
Colour: black
: Supplied with earthing stud.
: * Other colours and sizes are available on request. Please enquire for details.
- Catalog No:
- 115-0516
- DYCECZ06BM60045BK
- Colour:
- Grey
- Black