Order Entry
Finland
ContactUsLinkComponent
Splash heads, sloping, Quickfit®
Splash heads, sloping, Quickfit®
Catalog # BIBBSH7/43
Supplier:  SciLabware
undefined
Splash heads, sloping, Quickfit®
Catalog # BIBBSH7/43
Supplier:  SciLabware
Specifications
  • Cone to condenser:
    24/29 NS
  • Cone to flask:
    24/29 NS
  • Seq:
    06
  • Pk:
    5
Specifications
About this item
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Used in distillation assemblies to prevent raw liquid 'carrying over' from flask to condenser.