Distilling bridges, Claisen
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate 3.3 glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Still heads with Liebig condenser and Claisen attachment. With NS 14/23 socket for distillation thermometer, with 55 mm total length and glass vacuum tube connector. Connection for coolant water via tube connector.