Order Entry
Finland
Orders LinkContactUsLinkComponent
Distilling bridges
Distilling bridges
Supplier:   Lenz Laborglas
Distilling bridges
Supplier:  Lenz Laborglas
Certificates
Distilling bridges
Distilling bridges
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Still heads without condenser, with two vertical cones.
Documents
Product Family Options
Product Information
  • Jacket length
    Cone size
    Availability
    Price
  • 150 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 46,40 €
    46,40 €
    1 * 1 Items
       46,40 €
  • 200 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 63,90 €
    63,90 €
    1 * 1 Items
       63,90 €
  • 300 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 81,80 €
    81,80 €
    1 * 1 Items
       81,80 €
  • 350 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 94,00 €
    94,00 €
    1 * 1 Items
       94,00 €