Order Entry
Finland
ContactUsLinkComponent
Safety spectacles, Bandido
Safety spectacles, Bandido
Catalog # 111-0319
Supplier:  Bollé Safety
undefined
Safety spectacles, Bandido
Catalog # 111-0319
Supplier:  Bollé Safety
Specifications
  • Type:
    Anti-UV (2C-1,2), anti-scratch, anti-fog
  • Colour:
    Transparent
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Modern design safety glasses offering a good field of vision.
  • Wrap-around fit
  • Integrated side shields
  • ESP lens coating reduces glare and the harmful effects of blue light (only ESP model)
Weight: 28 g
EN 166, (EN 169, EN 170, EN 172)
Certifications: EN 166, (EN 169, EN 170, EN 172)