Order Entry
Finland
ContactUsLinkComponent
Safety spectacles, Viper
Safety spectacles, Viper
Catalog # 111-0340
Supplier:  Bollé Safety
undefined
Safety spectacles, Viper
Catalog # 111-0340
Supplier:  Bollé Safety
Specifications
  • Type:
    Anti-UV (3-1,2), anti-scratch
  • Colour:
    Dark blue
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
This model is ultra-light and very modern. With its one piece construction it provides additional protection and a wider field of vision.
  • Adjustable temple length
  • Integrated side shield for extra protection
Weight: 25 g
EN 166, EN 170
Certifications: EN 166, EN 170