Order Entry
Finland
ContactUsLinkComponent
Adhesive plate seal
Adhesive plate seal
Catalog # 732-0065
undefined
Adhesive plate seal
Catalog # 732-0065
Specifications
  • Description:
    Adhesive plate seal
  • Pk:
    100
Specifications
About this item
For long and short term storage of samples in microplates, and for sealing over pierced foil seals.
  • Non-adhesive tab for easy removal of seal
  • Temperature range −40 to +80 °C
  • Pierceable