Scalpel blades, non sterile
Supplier: BRESCO AB
Certificates
About this item
Stainless steel. Non sterile scalpel blades, individually packed in bundles of 10.
Models 10 to 15 fit handle No. 3, models 23 to 27 fit handle No. 4.
Stainless steel. Non sterile scalpel blades, individually packed in bundles of 10.
Models 10 to 15 fit handle No. 3, models 23 to 27 fit handle No. 4.