Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Retort Rings
Retort rings with bosshead
Retort rings with bosshead
Retort rings with bosshead
Supplier:
USBECK Laborgeräte
Certificates
Expand image
Retort rings with bosshead
Supplier:
USBECK Laborgeräte
About this item
Galvanised steel (8 mm Ø).
With integral bosshead for fixing to retort stand
Three-point bearing
Nickel plated brass screw