Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Racks
Tube Racks
Test tube racks
Test tube racks
:
KARTELL
Test tube racks
:
KARTELL
PP, white.
Excellent chemical resistance
Heat resistant up to 121 °C, autoclavable
Light, very robust
With two or three tiers.