Serological pipettes, Sterilin®
Supplier: Sterilin
Certificates
About this item
PS, transparent, disposable, sterile.
- Clear black printed graduations for maximum clarity, colour coded for easy identification, printed with product code/lot number and expiry date
- Negative graduations for extra capacity
- Gamma irradiated to ensure sterility and certified both non pyrogenic (<0,1 EU/ml) and non haemolytic
Ordering information: Batch specific certificate supplied in each case.
Caution: *Without suction adapter