Order Entry
Finland
Orders LinkContactUsLinkComponent
Microscope slides, Nunc™
   Sustainable Options Available
Microscope slides, Nunc™
   Sustainable Options Available
Certificates
Microscope slides, Nunc™
2 Options Available Below
Product Details & Documents
These microscope slides are designed for applications in cell culture.
The surface treatment ensures optimal conditions for cell attachment and growth.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2070
  • 734-2071
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • 734-2070
    Microscope slides, sterile, PS
    Microscope slides, sterile, PS
    25×75 mm
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 598,00 €
    598,00 €
    1 * 100 Items
       598,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2070
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 734-2071
    Microscope slides, sterile, Permanox™
    Microscope slides, sterile, Permanox™
    27×75 mm
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 498,00 €
    498,00 €
    1 * 100 Items
       498,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2071
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact