Cell culture dishes, CELLSTAR®
Supplier: Greiner Bio-One
Certificates
About this item
PS, cell culture-treated, sterile. Available with a variety of different dimensions and growth areas. The 57 cm² and 143 cm² dishes have an extra high profile, with a height of 20 mm. All dishes are certified free of RNase, DNase and human DNA, are non pyrogenic and non cytotoxic.
- Physical surface treatment for improved cell adhesion
- Vents ensure efficient gas exchange
- Easy stacking
Specifications
- Catalog No:
- 392-0370
- 391-3210
- 391-3205
- 391-3225
- 391-3236
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)