Hydrometers for petroleum
Supplier: Alla France
Certificates
About this item
These hydrometers for the petroleum industry have been designed with reading below the meniscus.
Certifications: ISO 649, NF 35511, BS 718, DIN 12791.
Ordering information: Traceable® three point calibration works certificate or official laboratory three point calibration certificate are available.
Specifications
- Catalog No:
- 614-0509
- 614-0500
- 614-0503
- 614-0506
- 614-0504
- 614-0501
- 614-0505
- 614-0502
- 614-0507
- 614-0508
- Length:
- 335 mm
- 335 mm
- 335 mm
- 335 mm
- 335 mm
- 335 mm
- 335 mm
- 335 mm
- 335 mm
- 335 mm