Order Entry
Finland
ContactUsLinkComponent
Goggles, Attack
Goggles, Attack
Catalog # 111-0313
Supplier:  Bollé Safety
undefined
Goggles, Attack
Catalog # 111-0313
Supplier:  Bollé Safety
Specifications
  • Type:
    Anti-scratch, anti-blur
  • Colour:
    Transparent
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
These goggles offer a panoramic vision and anti-scratch coating on both sides. They provide protection against impact, chemical splash, dust and liquid aerosols.
  • Cushioned frame provides maximum comfort and protection
  • Fitted with a wide, adjustable headband
  • Ideal for use with prescription spectacles due to the temple gaps
  • Can be used with certain respirators
Weight: 95 g
EN 166, EN 170
Certifications: EN 166, EN 170.