Cleanroom gloves, nitrile, Chemsoft CE™
These gloves have anti-static properties and are therefore ideal for the fine assembly of parts in cleanrooms, pharmaceutical areas and paints shop.
- Non sterile, powder-free
- Anti-static and state-of-the-art cleaning process reduces particulate contamination
- Ambidextrous and textured fingertips provide a good grip
- Rolled cuff prevents tearing while donning
: EN ISO 374-1 Type B (JKT); Cat. III; AQL 1,5
: Double bagged to ensure cleanliness.
- Catalog No:
- 112-2550
- 112-2547
- 112-2546
- 112-2549
- 112-2548
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1 Type B (JKT)
- EN ISO 374-1 Type B (JKT)
- EN ISO 374-1 Type B (JKT)
- EN ISO 374-1 Type B (JKT)
- EN ISO 374-1 Type B (JKT)
- Anti-static:
- +
- +
- +
- +
- +