Order Entry
Finland
Orders LinkContactUsLinkComponent
Vacuum filtration units
 :   Sartorius
Vacuum filtration units
 :  Sartorius
Vacuum filtration units
Vacuum filtration units
Borosilicate glass, aluminium clamp, autoclavable.
Vacuum filtration unit for clarification, particle testing and hybridisation tests.
516-7015: With glass frit filter support
511-0132: With PTFE-coated stainless steel screen filter support
  • Catalog No:
  • 516-7015
  • 511-0132
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Product Family Options
  • Capacity
    Material
    For filter Ø
  • 30 ml
    Borosilicate glass
    25 mm
    1 * 1 Items
    1 * 1 Items - 244,00 €
    244,00 €
    1 * 1 Items
       244,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    516-7015
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 30 ml
    Borosilicate glass
    25 mm
    1 * 1 Items
    1 * 1 Items - 360,00 €
    360,00 €
    1 * 1 Items
       360,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    511-0132
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
Accessories
  •  
    Description
  • SART6983008
    Small clamp
    1 * 1 Items - 178,00 €
  •  
    Description
  • SART6980117
    Silicone O-ring
    1 * 1 Items - 9,00 €
  •  
    Description
  • SART6980116
    Glass base
    1 * 1 Items - 71,00 €
  •  
    Description
  • SART6980115
    Glass funnel, 30 ml
    1 * 1 Items - 78,00 €
  •  
    Description
  • SART17149
    PTFE ring with PTFE coated underdrain screen and support screen including silicone O-ring
    1 * 1 Items - 170,00 €
  •  
    Description
  • SART17148
    PTFE ring with glass frit and silicone O-ring
    1 * 1 Items - 116,00 €